Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Flow rate:1,2 - 2,7 ml/min
- Particle Size:75 µm
- Capacity:4,7 ml
- Description:HiScreen™ Capto™ Phenyl (High Sub)
- Seq:01
- pH range:3 - 13
- Column hardware:PP
- Bed dimensions:7,7×100 mm
- Pk:1
Specifications
About this item
Capto™ Phenyl (high sub) and Capto™ Butyl are hydrophobic interaction chromatography (HIC) resins used in the capture and intermediate stages of protein purification.
- Excellent for method optimization and parameter screening due to the 10 cm bed height
- Easily connected in series to achieve 20 cm bed height
- Small bed volume for fast results and minimal sample/buffer consumption
- Reproducible results, scalable to BioProcess columns packed with the same resin using the same flow velocity
Capto™ Phenyl (high sub) is an excellent choice for initial and intermediate step purifications requiring a chromatography resin with resin to high hydrophobicity. Performance and selectivity are similar to that of Phenyl Sepharose™ 6 Fast Flow (high sub). Higher flow rates can be used with Capto™ Phenyl (high sub) during equilibration, wash and regeneration.