Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Capacity:1 ml
- Description:rProtein A Protein G GraviTrap™
- Seq:01
- Binding capacity:Approx. 35 mg human IgG/column
- Column hardware:PP
- Pk:10
rProtein A Protein G GraviTrap contains ten columns with 1 ml 50% rProtein A Sepharose Fast Flow and 50% Protein G Sepharose 4 Fast Flow resins.
- For general-purpose purification of monoclonal and polyclonal antibodies
- Suitable when the affinity for protein A and protein G is not known
- GraviTrap prepacked gravity flow columns enable simple one-step purification of proteins in 30 min without the need for equipment
- High purity: >95% in one step
- High protein recovery (typically 70% to 80%) due to high binding capacity of ~35 mg IgG/ml resin
This column can be used for manual purification of polyclonal and monoclonal antibodies from cell culture supernatant and biological fluids. It is particularly suitable when the affinity for protein A and protein G is not known. The gravity flow column relies on gravity and is reusable up to five times, so no additional instruments are needed for this protocol.