Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Flow rate:<4 ml/min
- Particle Size:34 µm
- Max. pressure:3 bar
- Capacity:1 ml
- Description:MAbTrap™ kit
- pH range:3 - 9
- Column hardware:PP
- Bed dimensions:7×25 mm
- Pk:1 KIT
Specifications
About this item
MAbTrap™ is an affinity chromatography kit for fast and effective purification of monoclonal and polyclonal IgG from ascites fluid, serum and cell culture supernatants. MAbTrap™ Kit contains everything necessary to perform a complete one step purification without using complicated equipment. It also eliminates time consuming buffer preparation and column packing.
- Purification of IgG from monoclonal and polyclonal material using a syringe in <15 minutes