About this item
HiTrap™ Octyl FF are prepacked Octyl Sepharose™ 4 Fast Flow columns for hydrophobic interaction chromatography (HIC).
- Provide convenient and fast small-scale protein purifications
- Flexible design for use with syringes, single laboratory pump, or chromatography system such as ÄKTA design
- High flow rates and high capacities
- Excellent and easy scale-up achieved by connecting several HiTrap™ columns in series or using HiPrep™ columns
The columns are used for small-scale protein purification as well as for screening of binding and elution conditions. Octyl Sepharose™ 4 Fast Flow is a well-established, standard aliphatic HIC medium for capture and intermediate purification.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Catalog No:
- GEHE17-5196-01
- GEHE17-1359-01
- Max. pressure:
- 5 bar
- 5 bar
- pH range:
- 3 - 13
- 3 - 13