Order Entry
China
ContactUsLinkComponent
Affinity chromatography media, IMAC Sepharose™ High Performance
Affinity chromatography media, IMAC Sepharose™ High Performance
目录编号 GEHE17-0920-07
供应商:  GE HEALTHCARE
CAS 编号:  
undefined
Affinity chromatography media, IMAC Sepharose™ High Performance
目录编号 GEHE17-0920-07
供应商:  GE HEALTHCARE
CAS 编号:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

规格

  • Particle Size:
    34 µm
  • Pk:
    100 ml
  • Description:
    IMAC Sepharose™ High Performance
  • pH range:
    3 - 12
  • Matrix:
    Highly cross-linked agarose, 6%
  • Storage temperature:
    4 - 30 °C
  • Binding capacity:
    At least 40 mg histidine-tagged protein
  • Flow rate:
    <300 cm/h
  • Type:
    Liquid

规格

关于此项目

IMAC Sepharose High Performance media is easily charged by the user with the metalion of choice for Immobilized Metal ion Affinity Chromatography (IMAC) and subsequent purification of polyhistidine tagged proteins.

  • For optimizing purification of histidine-tagged proteins when Ni2+ is not the best choice of metal ion
  • Conveniently charge with your metal of choice
  • 34 μm bead size for high performance purification
  • High binding capacity
  • Stable in commonly used aqueous buffers
  • Avoid using chelating agents, e.g. EDTA, EGTA, citrate

Conveniently charge with your metal of choice. IMAC Sepharose FF is supplied free of metal ions. It is charged by the user with the transition metal ion of choice (e.g. Cu₂₊, Zn₂₊, Ni₂₊, or Co₂₊); these metal ions will bind to the covalently immobilized chelating ligand on the Sepharose. Small bead size for high performance purification and high binding capacity.