Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Ø int.:7,70 mm
- Particle Size:90 µm
- Max. pressure:1,5 bar
- Capacity:4,7 ml
- Description:HiScreen™ Octyl FF
- Length:100 mm
- Seq:01
- pH range:3 - 13
- Column hardware:PP
- Pk:1
Specifications
About this item
HiScreen Octyl FF columns are prepacked with Octyl Sepharose 4 Fast Flow hydrophobic interaction chromatography (HIC) resins. The columns are an excellent choice for method optimization and parameter screening.
- Prepacked, ready-to-use columns for convenient process development optimization
- 10 cm bed height of HiScreen columns is designed to allow method optimization and parameter screening
- Easily connected in series to achieve 20 cm bed height
- Small bed volume gives fast results and minimal sample/buffer consumption
- Reproducible results, scalable to BioProcess columns packed with the same resin using the same linear fluid velocity
Octyl Sepharose 4 Fast Flow is a well-established, standard aliphatic HIC resin for capture and intermediate purification.
The small column volume of 4,7 ml and bed height of 10 cm make HiScreen columns excellent tools for method optimization, parameter screening, robustness testing, and convenient scale-up. Process fluid velocities can be applied, since the 10 cm bed height gives enough residence time and the results can then serve as basis for linear process scale-up.
If necessary, two columns can easily be connected in series to give a bed height of 20 cm.