Order Entry
China
Orders LinkContactUsLinkComponent
Affinity chromatography columns, HiTrap™ MabSelect™

Affinity chromatography columns, HiTrap™ MabSelect™

Supplier:   GE HEALTHCARE
Affinity chromatography columns, HiTrap™ MabSelect™
Supplier:  GE HEALTHCARE
Certificates
Affinity chromatography columns, HiTrap™ MabSelect™
Affinity chromatography columns, HiTrap™ MabSelect™

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

HiTrap™ MabSelect™ columns are prepacked with MabSelect™, a Bioprocess medium for capuring of monoclonal antibodies(Mabs) from large sample volumes.

  • Specificity of binding to the Fc region of IgG is similar to that of native protein A, which allows one-step purification
  • Oriented coupling of recombinant Protein A to the matrix via an engineered C-terminal cysteine enhances IgG binding capacity
  • Fewer regulatory concerns due to the total absence of mammalian culture in the ligand production and purification
Specifications for Product Family

Specifications

  • Catalog No:
  • GEHE28-4082-53
  • GEHE28-4082-55
  • GEHE28-4082-56
  • Max. pressure:
  • 5 bar
  • 5 bar
  • 5 bar
  • pH range:
  • 3 - 10
  • 3 - 10
  • 3 - 10
  • Storage:
  • 2 to 8 °C, 20% Ethanol (alternative storage solution 2% benzyl alcohol)
  • 2 to 8 °C, 20% Ethanol (alternative storage solution 2% benzyl alcohol)
  • 2 to 8 °C, 20% Ethanol (alternative storage solution 2% benzyl alcohol)

Specifications

Product Family Options

Product Information

  • Column hardware
    Particle Size
    Bed dimensions
    Capacity
    Flow rate
    Availability
    Price
  • GEHE28-4082-53
    HiTrap™ MabSelect™
    HiTrap™ MabSelect™
    PP
    85 µm
    7×25 mm
    5×1 ml
    <4 ml/min
    Log in for availability
    Log in for availability
    1 * 5 件
    1 * 5 件

    Specifications:
    More
    Specifications
    Cat. No.
    GEHE28-4082-53
    Max. pressure
    5 bar
    pH range
    3 - 10
    Storage
    2 to 8 °C, 20% Ethanol (alternative storage solution 2% benzyl alcohol)
  • GEHE28-4082-55
    HiTrap™ MabSelect™
    HiTrap™ MabSelect™
    PP
    85 µm
    16×25 mm
    1×5 ml
    <20 ml/min
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件

    Specifications:
    More
    Specifications
    Cat. No.
    GEHE28-4082-55
    Max. pressure
    5 bar
    pH range
    3 - 10
    Storage
    2 to 8 °C, 20% Ethanol (alternative storage solution 2% benzyl alcohol)
  • GEHE28-4082-56
    HiTrap™ MabSelect™
    HiTrap™ MabSelect™
    PP
    85 µm
    16×25 mm
    5×5 ml
    <20 ml/min
    Log in for availability
    Log in for availability
    1 * 5 件
    1 * 5 件

    Specifications:
    More
    Specifications
    Cat. No.
    GEHE28-4082-56
    Max. pressure
    5 bar
    pH range
    3 - 10
    Storage
    2 to 8 °C, 20% Ethanol (alternative storage solution 2% benzyl alcohol)

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.