Order Entry
China
ContactUsLinkComponent
Anion exchange chromatography columns, RESOURCE™ Q
Anion exchange chromatography columns, RESOURCE™ Q
Supplier:  GE HEALTHCARE
Certificates
undefined
Anion exchange chromatography columns, RESOURCE™ Q
Supplier:  GE HEALTHCARE

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Catalog No:
  • GEHE17-1179-01
  • GEHE17-1177-01
  • Description:
  • RESOURCE™ Q
  • RESOURCE™ Q
  • Max. pressure:
  • 6 bar
  • 15 bar
  • pH range:
  • 2 - 12
  • 2 - 12
  • Storage:
  • 4 to 30 °C, 20% Ethanol
  • 4 to 30 °C, 20% Ethanol

Specifications