Order Entry
China
ContactUsLinkComponent
Hydrophobic interaction chromatography media, Capto™ SP ImpRes
Hydrophobic interaction chromatography media, Capto™ SP ImpRes
Catalog # GEHE17-5468-02
Supplier:  GE HEALTHCARE
CAS Number:  
undefined
Hydrophobic interaction chromatography media, Capto™ SP ImpRes
Catalog # GEHE17-5468-02
Supplier:  GE HEALTHCARE
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Particle Size:
    36 - 44 µm
  • Pk:
    100 ml
  • Description:
    Capto™ SP ImpRes
  • Seq:
    01
  • Environmentally Preferable:
  • pH range:
    4 - 12
  • Matrix:
    High-flow agarose
  • Ligand:
    Sulfonate group
  • Storage temperature:
    4 - 30 °C
  • Binding capacity:
    >70 mg lysozyme, >95 mg BSA
  • Total ionic capacity:
    0,13 - 0,16 mmol H+/ml
  • Type:
    Liquid

Specifications

About this item

Capto™ SP ImpRes is a strong cation exchange bioprocessing medium, designed to meet the demands of modern large-scale manufacturers for fast, efficient, and cost-effective intermediate and polishing protein purification.

  • High resolution intermediate purification and polishing media based on the well established Capto™ platform with traditional ligands
  • Flexible process design due to large operational window of flow rates and bed heights
  • High throughput purifications easy to optimize and scale-up
  • Higher manufacturing productivity enables improved process economy
  • Bioprocessing media developed and supported for large scale chromatography

Capto SP ImpRes is a strong cation exchanger for the high-throughput intermediate purification and polishing steps of a wide range of biomolecules. This chromatography media is part of an expanded high-resolution platform based on the high-flow agarose Capto product line. By combining the high-flow characteristics of Capto media with a small particle size, Capto SP ImpRes delivers excellent pressure-flow properties with impressive resolution. The ability to run at higher flow rates and higher bed heights also increases flexibility in process design. Capto SP ImpRes is a BioProcess chromatography medium developed for large-scale purification of biopharmaceutical proteins. Based on the well-established Capto base matrix and with traditional ionic group ligands, it provides many opportunities for improved productivity and straightforward process development. Available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess media range, Capto SP ImpRes meets industrial demands with security of supply and comprehensive technical and regulatory support.