HiTrap Capto Q chromatography column is prepacked with a strong anion exchange modern resin for small scale protein purification.
- Suitable for screening of selectivity, binding and elution conditions, and for small-scale purifications
- Quaternary ammonium (Q) strong anion exchanger
- Optimized for capture and intermediate protein purification
- Convenient HiTrap format for easy connection to a syringe, peristaltic pump, or chromatography system such as an ÄKTA system
Capto Q resin couples a quaternary ammonium (Q) strong anion exchanger to a chemically modified, high-flow agarose matrix. The rigid agarose matrix provides greater particle rigidity without compromising pore size, which allows for high bed volume and purification of viscous samples at high flow rates.
High dynamic binding capacity is achieved with a dextran surface extender that coats the agarose matrix. The resin’s high binding-capacity, high flow rate, and low back pressure reduce process cycle times and improve productivity at process scale.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Catalog No:
- GEHE11-0013-03
- GEHE11-0013-02
- Max. pressure:
- 5 bar
- 5 bar
- pH range:
- 2 - 12
- 2 - 12
- Storage:
- 4...30 °C, 20% Ethanol
- 4...30 °C, 20% Ethanol