Affinity chromatography columns, HiTrap™ Protein L
About this item
Columns prepacked with Capto™ L, an affinity chromatography medium (resin) for capture of antibodies and antibody fragments such as Fab fragments, scFv, and Dabs.
- Prepacked for easy screening and process development, as well as small scale purification
- Columns easily connected in series for increased capacity
- Designed for use with syringe, peristalic pump, or chromatography system
3 Options Available Below
Product Details & Documents
Columns prepacked with Capto™ L, an affinity chromatography medium (resin) for capture of antibodies and antibody fragments such as Fab fragments, scFv, and Dabs.
- Prepacked for easy screening and process development, as well as small scale purification
- Columns easily connected in series for increased capacity
- Designed for use with syringe, peristalic pump, or chromatography system
Documents
Specifications for Product Family
Specifications
- Catalog No:
- GEHE17-5478-15
- GEHE17-5478-51
- GEHE17-5478-55
- Max. pressure:
- 5 bar
- 5 bar
- 5 bar
- pH range:
- 2 - 10
- 2 - 10
- 2 - 10
- Storage:
- 2 to 8 °C, 20% Ethanol
- 2 to 8 °C, 20% Ethanol
- 2 to 8 °C, 20% Ethanol
Specifications
Product Family Options
Product Information
- Column hardwareParticle SizeBed dimensionsCapacityFlow velocityAvailabilityPrice
- GEHE17-5478-15HiTrap™ Protein LSpecifications:
More
Specifications
Cat. No.GEHE17-5478-15Max. pressure5 barpH range2 - 10Storage2 to 8 °C, 20% Ethanol - GEHE17-5478-51HiTrap™ Protein LSpecifications:
More
Specifications
Cat. No.GEHE17-5478-51Max. pressure5 barpH range2 - 10Storage2 to 8 °C, 20% Ethanol - GEHE17-5478-55HiTrap™ Protein LSpecifications:
More
Specifications
Cat. No.GEHE17-5478-55Max. pressure5 barpH range2 - 10Storage2 to 8 °C, 20% Ethanol
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



