About this item
HiTrap™ CM FF is pre-packed with CM Sepharose™ Fast Flow, and is a weak cation exchanger for small scale protein purifications, as well as screening of binding and elutions conditions.
- Fast, easy ion exchange separations either alone or connected in series
- High flow rates and good scale-up potential
- Use a weak ion exchanger if the selectivity of a strong ion exchanger is insufficient
- Predictable scale-up
Q, SP, DEAE adn CM Sepharose Fast Flow are based on a robust, 6% highly cross-linked beaded agarose matrix with good flow properties and high loading capacities. ANX Sepharose 4 Fast Flow (high sub) is based on 4% highly cross-linked beaded agarose. This results in a medium with higher porosity, which is particularly useful for the purification of high molecular mass proteins.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Catalog No:
- GEHE17-5056-01
- GEHE17-5155-01
- Max. pressure:
- 5 bar
- 5 bar
- pH range:
- 4 - 13
- 4 - 13
Specifications
Related Information
17505601_DATA FILE.pdf
Datasheet
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements