About this item
HiTrap Q FF is prepacked with Q Sepharose Fast Flow, and is a strong anion exchanger for small-scale protein purifications as well as screening of binding and elutions conditions.
- Convenient and affordable for fast, easy ion exchange separations either alone or connected in series
- The industry standard for ion exchange chromatography
- High flow rates and good scale-up potential
- Use a weak ion exchanger if the selectivity of a strong ion exchanger is insufficient
- Predictable scale-up
Composed of crosslinked 6% agarose beads with quaternary amine strong anion exchange groups, this resin has high chemical stability, allowing well proven cleaning-in-place (CIP) and sanitization protocols. The resin’s 90 µm bead size yields rapid flow rates, making it simple to scale up the purification.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Catalog No:
- GEHE17-5053-01
- GEHE17-5156-01
- Max. pressure:
- 5 bar
- 5 bar
- pH range:
- 2 - 12
- 2 - 12