Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Capacity:5,3 ml
- Description:PD MidiTrap™ G-10
- Column hardware:PP
- Bed dimensions:13×40 mm
- Pk:50
Specifications
About this item
PD MidiTrap G-10 columns are prepacked, single-use gravity columns for buffer exchange and cleanup of biological samples, for example peptides, small proteins, and oligosaccharides.
- Easy cleanup of peptides, small proteins, or carbohydrates larger than Mr 700, before downstream analysis such as liquid chromatography, electrophoresis, and MS
- Efficient removal of contaminants such as salts, dyes, and radioactive labels
- High desalting capacity
- Preparation of samples in the volume range 400 to 1000 μl
PD MidiTrap G-10 gravity columns give fast and simple desalting and buffer exchange without any need for a purification system, PD MiniTrap G-10 columns are prepacked and designed for rapid, convenient sample clean-up of small proteins, peptides, oligo-saccharides and other small biomolecules (>700 Mr).
For smaller scale please use PD MiniTrap G-10 columns (300 μl).