Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:PD SpinTrap™ G-25
- Storage:4 to 30 °C, 0,05% Kathon CG/ICP Biocide
- Pk:50
- Sample volume:100 - 180 μl ml
Specifications
About this item
PD SpinTrap G-25 is a single-use column for cleanup of large biomolecules.
- Convenient and fast small-scale cleanup of proteins and other large biomolecules (>5000 Mr) with the possibility to run multiple samples in parallel
- Range of applications such as desalting, buffer exchange, and removal of low-molecular weight compounds
- High desalting capacity
- Reproducible, high throughput, parallel cleanup of protein samples
- Sample preparation in the range of 100 to 180 μl without sample dilution
PD SpinTrap G-25 is a single-use microspin column that is designed for desalting and buffer exchange of biological sample using a standard microcentrifuge. The column is prepacked with ~600 µL Sephadex G-25 resin, and the column material is biocompatible polypropylene.