About this item
HiPrep Sephacryl S-100 HR are prepacked gel filtration columns for preparative separation of biomolecules from up to 13 ml sample volume.
- Maximum recovery and minimum nonspecific adsorption with long-term chemical and physical stability resulting from the hydrophilic, rigid allyl dextran/bisacrylamide matrix
- Gel filtration columns for low to medium pressure ÄKTA systems.
- Well-established in industrial production
- The HiPrep Sephacryl series offers reproducible separations over a wide molecular weight range
- Convenient, prepacked HiPrep Sephacryl columns speed up protein purification
Sephacryl S-100 is used for separating peptides and small proteins.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Catalog No:
- GEHE17-1165-01
- GEHE17-1194-01
- Max. pressure:
- 1,5 bar
- 1,5 bar
- pH range:
- 3 - 11
- 3 - 11
- Storage:
- 4 to 30 °C, 20% Ethanol
- 4 to 30 °C, 20% Ethanol