Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Particle Size:90 µm
- Pk:15 g
- Description:CNBr-activated Sepharose™ 4B
- Seq:01
- pH range:2 - 11
- Matrix:Agarose, 4%
- Storage temperature:2 - 8 °C
- Binding capacity:25 mg α-chymotrypsinogen/ml medium-60 mg α-chymotrypsinogen/ml
- Type:Solid
Specifications
About this item
CNBr-activated Sepharose™ 4B is pre-activated media used for coupling antibodies or other large proteins containing -NH₂ groups to the Sepharose™ media, by the cyanogen bromide method, without an intermediate spacer arm.
- Multipoint attachment resulting in a chemically stable product
- Over 25 years proven track record for coupling ligands containing primary amino groups
- The coupling reaction by the cyanogen bromide method is spontaneous, rapid and easy to carry out
- Stable to all commonly used aqueous solutions
- Can be used with non-ionic detergents, denaturing solvents, e.g. 8 M urea and 6 M guanidine hydrochloride
- Stable in organic solvents, such as 50% dimethylformamide and 50% dioxane
It provides a very convenient way to immobilise ligands by the cyanogen bromide method. The coupling reaction is spontaneous, rapid and easy to carry out. The application area covers immobilisation of proteins, peptides and nucleic acids. For immobilising antibodies or other large proteins containing -NH₂ groups by coupling them to the matrix.