Bis(tri-tert-butylphosphine)palladium(0)
Catalog # CHLCR01661280141
Supplier: LOCAL DEALS
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
:
:511.05 g/mol
CAS Number:53199-31-8
Formula:C₂₄H₅₄P₂Pd
MDL Number:MFCD03094580