Tri-tert-butylphosphonium tetrafluoroborate
Catalog # CHLCR01434420144
Supplier: LOCAL DEALS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:290.13 g/mol
CAS Number:131274-22-1
:MFCD07189501
Formula:C₁₂H₂₈BF₄P
MDL Number:MFCD04039975