Hexamethylphosphoramide
Catalog # CHLCR00411730121
Supplier: LOCAL DEALS
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 ml
- Seq:2440
Specifications
About this item
:
EINECS:211-653-8
CAS Number:680-31-9
Formula:C₆H₁₈N₃OP
MDL Number:MFCD00008303