
Lithium chloride
Catalog # CHLCR00932310111
Supplier: LOCAL DEALS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:42.39 g/mol
Merck Index:13,05550
Boiling Pt:1382 °C (1013 hPa)
Melting Pt:614 °C
Density:2.07 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-212-3
:GHS07
CAS Number:7447-41-8
Formula:LiCl
MDL Number:MFCD00011078