Cyclohexanone
Glass bottle, 500 ml
CHLCR01947120124
: Pimelic Ketone
- :108-94-1
- :C₆H₁₀(=O)
- :MFCD00001625
- :98.14 g/mol
- :13,02755
- :155.6 °C (1013 hPa)
- :–31 °C
- :0.9478 g/cm³ (20 °C)
- :43 °C
- :Ambient
- :203-631-1
- :1915
- :3,III
- :01-2119453616-35
- Pack type:Glass bottle
- Pk:500 ml