
Sodium benzoate dense 99.01-101.0% (by anhydrous basis) NF, FCC, Macron Fine Chemicals™
Catalog # MACR1331-26
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Fibre drum
- Pk:45 kg
- Seq:4388
Specifications
Specification Test Results
- NF - Assay (C₆H₅COONa) (anhydrous basis):99.0 - 101.0 %
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- NF - Identification C:Passes Test
- NF - Alkalinity:Passes Test
- NF - Water (H₂O):≤1.5 %
- FCC - Assay (C₇H₅NaO₂) (anhydrous basis):99.0 - 101.0 %
- FCC - Alkalinity (as NaOH):≤0.04 %
- FCC - Arsenic (As):≤3 ppm
- FCC - Identification A:Passes Test
- FCC - Identification B:Passes Test
- FCC - Identification C:Passes Test
- FCC - Lead (Pb):≤2 mg/kg
- FCC - Water (H₂O):≤1.5 %
- Solubility:Passes Test
- Appearance (white, crystalline granules):Passes Test
Specification Test Results
About this item
:
:144.11 g/mol
Merck Index:13,08654
Melting Pt:410…430 °C
Density:1.44 g/cm³ (20 °C)
Flash Pt:>100 °C
EINECS:208-534-8
REACH:01-2119460683-35
:GHS07
Storage Temperature:Ambient
CAS Number:532-32-1
Formula:C₆H₅COONa
MDL Number:MFCD00012463