
4-Bromophenol
Supplier: LOCAL DEALS
Certificates
About this item
CAS Number:106-41-2
:
Formula:C₆H₅BrO
MDL Number:MFCD00002313
:173.01 g/mol
Merck Index:13,01413
Boiling Pt:236 °C (1013 hPa)
Melting Pt:63 °C
Density:∼1.84 g/cm³ (20 °C)
Flash Pt:>110 °C
Storage Temperature:Ambient
EINECS:203-394-4
UN:2811
ADR:6.1,III
:GHS07