

Dichloromethane
Catalog # CHLC6603-04
Supplier: LOCAL DEALS
CAS Number:
Specifications
- Pk:4 L
- Seq:5162
Specifications
About this item
:
:84.93 g/mol
Merck Index:12,06140
Boiling Pt:39.8 °C (1013 hPa)
Melting Pt:–95 °C
Density:1.322 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-838-9
UN:1593
ADR:6.1,III
:GHS08
:GHS07
CAS Number:75-09-2
Formula:CH₂Cl₂
MDL Number:MFCD00000881