


1,2-Dichloroethane
Catalog # CHLCR00273220024
Supplier: LOCAL DEALS
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 ml
- Seq:2725
Specifications
About this item
:
:98.96 g/mol
Merck Index:13,03831
Boiling Pt:83.5 °C (1013 hPa)
Melting Pt:–35.5 °C
Density:1.254 g/cm³ (20 °C)
Flash Pt:13 °C
EINECS:203-458-1
UN:1184
ADR:3,II
:GHS02
:GHS08
:GHS07
Storage Temperature:Ambient
CAS Number:107-06-2
Formula:ClH₂CCH₂Cl
MDL Number:MFCD00000963