Order Entry
China
Orders LinkContactUsLinkComponent
Formic acid

Formic acid

Flammable
Corrosive
Supplier:   LOCAL DEALS
Formic acid
Supplier:  LOCAL DEALS
Certificates
Formic acid

2 Options Available Below

Product Details & Documents
Synonyms:  Aminic Acid, Methanoic acid
  • CAS Number:64-18-6
  • Formula:HCO₂H
  • MDL Number:MFCD00003297
  • Molecular Weight:46.03 g/mol
  • Boiling Pt:100…101 °C (1013 hPa)
  • Melting Pt:8.2…8.4 °C
  • Density:1.22 g/cm³ (25 °C)
  • Flash Pt:48 °C
  • EINECS:200-579-1
  • ADR:8,II
  •  :Density: 1.22 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • UN:1779
  • REACH:01-2119491174-37
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 毫升
    1 * 250 毫升

  • Glass bottle
    500 ml
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升