Butyl acetate
Glass bottle, 500 mL
Catalog # CHLCR01943520124
Supplier: LOCAL DEALS
Jump to:
Product Details & Documents
Synonyms: Butyl ethanoate, Acetic acid butyl ester, Acetic acid n-butyl ester, n-Butyl acetate
- CAS Number:123-86-4
- Formula:CH₃COO(CH₂)₃CH₃
- MDL Number:MFCD00009445
- Molecular Weight:116.16 g/mol
- Merck Index:13,01534
- Boiling Pt:126 °C (1013 hPa)
- Melting Pt:–76 °C
- Density:0.875 g/cm³ (20 °C)
- Flash Pt:25 °C
- Storage Temperature:Ambient
- EINECS:204-658-1
- UN:1123
- ADR:3,III
Specifications
Specifications
- Pack type:Glass bottle
- Pk:500 mL



