Lithium chloride monohydrate
Glass bottle for solids, 500 g
Catalog # CHLCR00227720124
Supplier: LOCAL DEALS
Jump to:
Product Details & Documents
Synonyms: Lithium chloride hydrate, Hydrochloric acid lithium salt monohydrate
- CAS Number:16712-20-2
- Formula:ClLi·1H₂O
- MDL Number:MFCD00149764
- Molecular Weight:60.41 g/mol
- Storage Temperature:Ambient
- EINECS:231-212-3
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



