Order Entry
China
ContactUsLinkComponent
Sodium hydroxide 0.1 mol/l (0.1 N) in aqueous solution
Sodium hydroxide 0.1 mol/l (0.1 N) in aqueous solution
Catalog # CHLCR00275070024
Supplier:  LOCAL DEALS
CAS Number:  
Sodium hydroxide 0.1 mol/l (0.1 N) in aqueous solution
Catalog # CHLCR00275070024
Supplier:  LOCAL DEALS
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 ml
  • Seq:
    2725
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Caustic soda
 :
 :40 g/mol
Density:1 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,III
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548