



Hexane (mixture of isomers)
Catalog # HONEAH216-4
Supplier: HONEYWELL SPECIALTY CHEMICALS
CAS Number:
Specifications
- Pk:4 L
- Seq:5162
Specifications
About this item
:
:86.18 g/mol
Merck Index:13,04712
Boiling Pt:68…70 °C (1013 hPa)
Melting Pt:–95 °C
Density:0.66 g/cm³ (20 °C)
Flash Pt:–22 °C
Storage Temperature:Ambient
EINECS:203-777-6
UN:1208
ADR:3,II
:GHS02
:GHS08
:GHS07
:GHS09
CAS Number:110-54-3
:92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD02179311