L-Phenylalanine 98.5-101.5% (dried basis) USP, Multi-Compendial, J.T.Baker®
: Avantor
:63-91-2
:
:C₉H₁₁NO₂
:MFCD00064227
:165.19 g/mol
:14,07271
:313 °C (1013 hPa)
:173 °C
:0.754 g/cm³ (20 °C)
:Ambient
:200-568-1
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Meets B.P. Chemical Specifications:98.5 - 101.5%
- Meets E.P. Chemical Specifications:Passes Test
- GMP Manufactured Product:-34.7 - -32.7 Degree
- Meets U.S.P Requirements:5.4 - 6.0
- Meets J.P. Chemical Specifications:≤0.3%
- Preserve in well-closed containers.:≤0.4%
- Store in well-closed,light-resistant containers:≤0.05%
- CAUTION: For Manufacturing, processing or repackaging:≤0.03%
- Bulk Pharmaceutical Chemical:≤0.003%
- USP - Assay (C₉H₁₁NO₂) (dried basis):98.5 - 101.5 %
- USP - Identification:Passes Test
- USP - Specific Rotation [Å]²⁵ ^D (+):-34.7 - -32.7 Degree
- USP - pH:5.4 - 6.0
- USP - Loss on Drying at 105°C:≤0.3 %
- USP - Residue on Ignition:≤0.4 %
- USP - Chloride (Cl):≤0.05 %
- USP - Sulfate (SO₄):≤0.03 %
- USP - Iron (Fe):≤0.003 %
- USP - Related Compounds - Individual Impurities:≤0.5 %
- USP - Related Compounds - Total Impurities:≤2.0 %
- EP/BP - Assay (C₉H₁₁NO₂) (dried basis):98.5 - 101.0 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Specific Rotation [Å]²º^D (+):-35.5 - -33.0 Degree
- EP/BP-Ninhydrin-Positive Substances-Each:≤0.2 %
- EP/BP-Ninhydrin-Positive Substances-Total Impurities:≤0.5 %
- EP/BP - Chloride (Cl):≤200 ppm
- EP/BP - Sulfate (SO₄):≤300 ppm
- EP/BP - Ammonium (NH₄):≤0.02 %
- EP/BP - Iron (Fe):≤10 ppm
- EP/BP - Loss on Drying:≤0.5 %
- EP/BP - Ash (sulfated):≤0.1 %
- JP - Assay (C₉H₁₁NO₂) (dried basis):98.5 - 101.0 %
- JP - Identification:Passes Test
- JP - Optical Rotation (+):-35.5 - -33.0 Degree
- JP - pH:5.3 - 6.3
- JP - Clarity and Color of Solution:Passes Test
- JP - Chloride (Cl):≤0.021 %
- JP - Sulfate (SO₄):≤0.028 %
- JP - Ammonium (NH₄):≤0.02 %
- JP - Heavy Metals (as Pb):≤20 ppm
- JP - Arsenic (As):≤2 ppm
- JP - Related Substances:Passes Test
- JP - Loss on Drying at 105°C:≤0.30 %
- JP - Residue on Ignition:≤0.10 %