Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
¥0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
China
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Di-tert-butyl peroxide
Di-tert-butyl peroxide
Di-tert-butyl peroxide
CHLCR00182230123
:
LOCAL DEALS
:
Di-tert-butyl peroxide
CHLCR00182230123
:
LOCAL DEALS
:
Pack type:
Glass bottle
Pk:
250 ml
Seq:
2590
:
:
146.23 g/mol
:
12,03515
:
109 °C (1013 hPa)
:
< –25 °C
:
0.798 g/cm³ (20 °C)
:
12 °C
:
203-733-6
:
GHS02
:
110-05-4
:
C₈H₁₈O₂
:
MFCD00008803