Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
¥0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
China
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
n-Butyllithium
n-Butyllithium 1.6 mol/l in hexane
n-Butyllithium 1.6 mol/l in hexane
n-Butyllithium 1.6 mol/l in hexane
Supplier:
LOCAL DEALS
Certificates
Expand image
n-Butyllithium 1.6 mol/l in hexane
Supplier:
LOCAL DEALS
About this item
CAS Number:
109-72-8
:
Formula:
C₄H₉Li
MDL Number:
MFCD00009414
:
64.06 g/mol
Density:
0.69 g/cm³ (20 °C)
Flash Pt:
–22 °C
EINECS:
203-698-7
UN:
3394
ADR:
4.2,I