Order Entry
China
ContactUsLinkComponent
Sodium hypochlorite (5% Cl₂) in aqueous solution, Laboratory Reagent, sterile, J.T.Baker®
Corrosive
Environment
Sodium hypochlorite (5% Cl₂) in aqueous solution, Laboratory Reagent, sterile, J.T.Baker®
Catalog # BAKRP005-03
Supplier:  Avantor
CAS Number:  
undefined
Corrosive
Environment
Sodium hypochlorite (5% Cl₂) in aqueous solution, Laboratory Reagent, sterile, J.T.Baker®
Catalog # BAKRP005-03
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    1 gal US
  • Seq:
    8080

Specifications

Specification Test Results

  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Assay (NaClO):
    4.0 - 6.0 %
  • Mercury (Hg):
    ≤5.0 ppb
  • The above referenced lot has passed the current USP Test for Sterility.:
    = 5.0 ppb

Specification Test Results

About this item

 :
 :74.44 g/mol
Boiling Pt:111 °C (1013 hPa)
Melting Pt:–30…–20 °C
Density:1.22…1.25 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-668-3
UN:1791
ADR:8,II
 :Management of Change (MOC) category = NI
 :GHS05
 :GHS09
CAS Number:7681-52-9
Formula:NaOCl
MDL Number:MFCD00011120