

Sodium hypochlorite (10% Cl₂) in aqueous solution
Catalog # CHLCR00389290115
Supplier: LOCAL DEALS
CAS Number:
Specifications
- Pk:10 kg
- Seq:4158
Specifications
About this item
:
:74.44 g/mol
Boiling Pt:111 °C (1013 hPa)
Melting Pt:–30…–20 °C
Density:1.22…1.25 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:231-668-3
UN:1791
ADR:8,II
:GHS05
:GHS09
CAS Number:7681-52-9
Formula:ClNaO
MDL Number:MFCD00011120