Order Entry
China
ContactUsLinkComponent
Sodium hypochlorite (10% Cl₂) in aqueous solution
Corrosive
Environment
Sodium hypochlorite (10% Cl₂) in aqueous solution
Catalog # CHLCR00389220124
Supplier:  LOCAL DEALS
CAS Number:  
Corrosive
Environment
Sodium hypochlorite (10% Cl₂) in aqueous solution
Catalog # CHLCR00389220124
Supplier:  LOCAL DEALS
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :74.44 g/mol
Boiling Pt:111 °C (1013 hPa)
Melting Pt:–30…–20 °C
Density:1.22…1.25 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:231-668-3
UN:1791
ADR:8,II
 :GHS05
 :GHS09
CAS Number:7681-52-9
Formula:ClNaO
MDL Number:MFCD00011120