Order Entry
China
ContactUsLinkComponent
Sodium chloride 0,9% in aqueous solution
Sodium chloride 0,9% in aqueous solution
Catalog # CHLCR02112370124
Supplier:  LOCAL DEALS
CAS Number:  
Sodium chloride 0,9% in aqueous solution
Catalog # CHLCR02112370124
Supplier:  LOCAL DEALS
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 ml
  • Seq:
    2725
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Saline, NaCl, Sea salt, Halite
 :
 :58.44 g/mol
Storage Temperature:Ambient
EINECS:231-598-3
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477