Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
¥0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
China
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 4-fluorobenzoate
Methyl-4-fluorobenzoate
Methyl-4-fluorobenzoate
Methyl-4-fluorobenzoate
CHLCR01608025133
:
LOCAL DEALS
:
Methyl-4-fluorobenzoate
CHLCR01608025133
:
LOCAL DEALS
:
Pack type:
Glass bottle for solids
Pk:
25 g
Seq:
3308
:
:
154.14 g/mol
:
206-956-7
:
403-33-8
:
C₈H₇FO₂
:
MFCD00017959