Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
¥0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
China
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl 4-fluorobenzoate
Ethyl-4-fluorobenzoate
Ethyl-4-fluorobenzoate
CHLCSY003268
:
LOCAL DEALS
:
Ethyl-4-fluorobenzoate
CHLCSY003268
:
LOCAL DEALS
:
Pack type:
Glass bottle for solids
Pk:
50 g
Seq:
3368
:
:
168.17 g/mol
:
13,04198
:
210 °C (1013 hPa)
:
26 °C
:
1.146 g/cm³ (20 °C)
:
207-194-8
:
451-46-7
:
C₉H₉FO₂
:
MFCD00000351