Order Entry
China
ContactUsLinkComponent
Methyl-4-bromobenzoate
  CHLCR01515730121
 :  
Methyl-4-bromobenzoate
  CHLCR01515730121
 :  
  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401
 :
 :215.05 g/mol
 :13,01392
 :246 °C (1013 hPa)
 :78…81 °C
 :1.689 g/cm³ (20 °C)
 :210-596-6
 :619-42-1
 :C₈H₇BrO₂
 :MFCD00013531