


Ethylene glycol monomethyl ether (2-methoxyethanol)
Catalog # CHLCR01103330124
Supplier: LOCAL DEALS
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 ml
- Seq:2725
Specifications
About this item
:
:76.1 g/mol
Merck Index:12,06118
Boiling Pt:124.6 °C (1013 hPa)
Melting Pt:–85 °C
Density:0.964 g/cm³ (20 °C)
Flash Pt:39 °C
EINECS:203-713-7
UN:1188
ADR:3,III
:GHS02
:GHS08
:GHS07
Storage Temperature:Ambient
CAS Number:109-86-4
Formula:C₃H₈O₂
MDL Number:MFCD00002867