

Diisopropyl ether
Catalog # CHLCR01102030124
Supplier: LOCAL DEALS
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 ml
- Seq:2725
Specifications
About this item
:
:102.18 g/mol
Merck Index:13,05232
Boiling Pt:68.5 °C (1013 hPa)
Melting Pt:–86 °C
Density:0.726 g/cm³ (20 °C)
Flash Pt:–28 °C
Storage Temperature:Ambient
EINECS:203-560-6
UN:1159
ADR:3,II
:GHS02
:GHS07
CAS Number:108-20-3
Formula:(CH₃)₂CHOCH(CH₃)₂
MDL Number:MFCD00008880