

Diisopropyl ether
Supplier: LOCAL DEALS
Certificates
About this item
CAS Number:108-20-3
:
Formula:(CH₃)₂CHOCH(CH₃)₂
MDL Number:MFCD00008880
:102.18 g/mol
Merck Index:13,05232
Boiling Pt:68.5 °C (1013 hPa)
Melting Pt:–86 °C
Density:0.726 g/cm³ (20 °C)
Flash Pt:–28 °C
Storage Temperature:Ambient
EINECS:203-560-6
UN:1159
ADR:3,II
:GHS02
:GHS07