

Bis(2-methoxyethyl)ether
Catalog # CHLCR00142930124
Supplier: LOCAL DEALS
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
About this item
:
Merck Index:13,03187
Boiling Pt:162 °C (1013 hPa)
Melting Pt:–64 °C
Density:0.944 g/cm³ (20 °C)
Flash Pt:53 °C
Storage Temperature:Ambient
EINECS:203-924-4
UN:3271
ADR:3,III
:134.18 g/mol
:GHS02
:GHS08
CAS Number:111-96-6
Formula:C₆H₁₄O₃
MDL Number:MFCD00008503