酒石酸钾钠四水合物
Catalog # CHLCR00437120124
Supplier: LOCAL DEALS
CAS Number:
Specifications
- 包装类型:Glass bottle for solids
- 包装规格:500 g
- 顺序:3645
Specifications
About this item
:
:282.22 g/mol
Merck Index:13,07755
Melting Pt:70…80 °C
Density:∼1,767 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:206-156-8
CAS Number:6381-59-5
Formula:KOOC(CHOH)₂COONa·4H₂O
MDL Number:MFCD00150989