

(±)-2-Bromopropionic acid
Catalog # CHLCR00251430133
Supplier: LOCAL DEALS
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mL
- Seq:2315
Specifications
About this item
:
:152.98 g/mol
Boiling Pt:203 °C (1013 hPa)
Melting Pt:∼25 °C
Density:1.7 g/cm³ (25 °C)
Flash Pt:100 °C
EINECS:209-947-6
UN:3261
ADR:8,II
:GHS05
:GHS07
CAS Number:598-72-1
Formula:H₃CBrCHCOOH
MDL Number:MFCD00004211