
Iron(II) sulphate heptahydrate
Catalog # CHLCR02406420124
Supplier: LOCAL DEALS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:278.02 g/mol
Merck Index:13,04091
Melting Pt:64 °C
Density:1.9 g/cm³ (20 °C)
EINECS:231-753-5
:GHS07
Storage Temperature:Ambient
CAS Number:7782-63-0
Formula:FeSO₄·7H₂O
MDL Number:MFCD00149719