

tert-Butyl methyl ether
Catalog # CHLCR00334320124
Supplier: LOCAL DEALS
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 ml
- Seq:2725
Specifications
About this item
:
:88.15 g/mol
Merck Index:13,06059
Boiling Pt:55.2 °C (1013 hPa)
Melting Pt:–108.6 °C
Density:0.7405 g/cm³ (20 °C)
Flash Pt:–28 °C
Storage Temperature:Ambient
EINECS:216-653-1
UN:2398
ADR:3,II
:GHS02
:GHS07
CAS Number:1634-04-4
Formula:(CH₃)₃COCH₃
MDL Number:MFCD00008812