tert-Butyl methyl ether
20 kg
Catalog # CHLCR00334300100
Supplier: LOCAL DEALS
Jump to:
Product Details & Documents
Synonyms: MTBE, 2-Methoxy-2-methylpropane, Methyl-t-Butyl Ether
- CAS Number:1634-04-4
- Formula:(CH₃)₃COCH₃
- MDL Number:MFCD00008812
- Molecular Weight:88.15 g/mol
- Merck Index:13,06059
- Boiling Pt:55.2 °C (1013 hPa)
- Melting Pt:–108.6 °C
- Density:0.7405 g/cm³ (20 °C)
- Flash Pt:–28 °C
- Storage Temperature:Ambient
- EINECS:216-653-1
- UN:2398
- ADR:3,II
Specifications
Specifications
- Pk:20 kg



